missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD1 Antibody (OTI10D8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74005B
This item is not returnable.
View return policy
Description
SCFD1 Monoclonal specifically detects SCFD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SCFD1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| SCFD1 | |
| Recombinant protein fragment corresponding to amino acids 66-190 of human SCFD1 (NP_878255) produced in E.coli. | |
| 0.1 mL | |
| 23256.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI10D8 | |
| Western Blot, Immunohistochemistry | |
| C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction