missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97778-100ul
This item is not returnable.
View return policy
Description
SCFD1 Polyclonal antibody specifically detects SCFD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SCFD1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| C14orf163, KIAA0917chromosome 14 open reading frame 163, sec1 family domain containing 1, sec1 family domain-containing protein 1, SLY1, SLY1 homolog, sly1p, STXBP1L2RA410, Syntaxin-binding protein 1-like 2, vesicle transport-related protein | |
| Produced in rabbits immunized with E. coli-derived Human SCFD1 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 23256 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction