Learn More
PeproTech SCF Polyclonal Antibody, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Brand: PeproTech 500-P48G-100UG
Description
AA Sequence of recombinant protein: MEGICRNRVT NNVKDVTKLV ANLPKDYMIT LKYVPGMDVL PSHCWISEMV VQLSDSLTDL LDKFSNISEG LSNYSIIDKL VNIVDDLVEC VKENSSKDLK KSFKSPEPRL FTPEEFFRIF NRSIDAFKDF VVASETSDCV VSSTLSPEKD SRVSVTKPFM LPPVA. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human SCF. Anti-Human SCF-specific antibody was purified by affinity chromatography employing an immobilized Human SCF matrix. Sandwich ELISA: To detect Human SCF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human SCF (500-P48GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human SCF. Western Blot: To detect hSCF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hSCF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
Specifications
| SCF | |
| Polyclonal | |
| Unconjugated | |
| KITLG | |
| 710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 4254 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P21583 | |
| KITLG | |
| E.coli-derived Recombinant Human SCF. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.