missing translation for 'onlineSavingsMsg'
Learn More

PeproTech SCF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

£177.00 - £2745.00

Specifications

Antigen SCF
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17843303
View Documents
PeproTech
500-P71BT-25UG
25 μg
£177.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17823043
View Documents
PeproTech
500-P71BT-50UG
50 μg
£228.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17803673
View Documents
Invitrogen
500-P71BT-1MG
2 x 500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30054406
View Documents
PeproTech
500-P71BT-500UG
500 μg
£1466.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: MKEICGNPVT DNVKDITKLV ANLPNDYMIT LNYVAGMDVL PSHCWLRDMV IQLSLSLTTL LDKFSNISEG LSNYSIIDKL GKIVDDLVLC MEENAPKNIK ESPKRPETRS FTPEEFFSIF NRSIDAFKDF MVASDTSDCV LSSTLGPEKD SRVSVTKPFM LPPVA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine SCF. Anti-Murine SCF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mSCF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine SCF (500-P71) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mSCF. Western Blot: To detect mSCF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mSCF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P71BT-1MG will be provided as 2 x 500 μg

Stem cell factor (SCF) is the ligand of the c-Kit oncogene and is expressed by various structural and inflammatory cells in the airways. Binding of SCF by the c-Kit receptor leads to homodimerization of the receptor and the activation of signalling pathways such as PI-3, PLC-gamma, Jak/STAT, and MAP kinase pathways. SCF expression leads to the induction of mast cell survival and the expression and release of histamine, pro-inflammatory cytokines and chemokines. The inhibition of the SCF/c-Kit pathway leads to a decrease in histamine levels, mast cell and eosinophil infiltration, IL-4 production and airway hyperresponsiveness, suggesting this pathway may be a useful therapeutic target in inflammatory diseases such as asthma. At least two isoforms of SCF are known to exist.
Specifications

Specifications

SCF
ELISA, Western Blot
Biotin
Rabbit
Mouse
P20826
17311
E.coli-derived, 18.3 kDa Recombinant Murine SCF.
Antigen affinity chromatography
Kitl
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
710-712; blz; c-Kit ligand; Clo; cloud gray; Con; contrasted; CSF; DCUA; DFNA69; DKFZp686F2250; familial progressive hyperpigmentation 2; FPH2; FPHH; Gb; grizzle-belly; hematopoietic growth factor KL; H-SCF; Kit ligand; Kitl; Kitlg; kit-ligand; KL-1; Mast cell growth factor; MGF; M-SCF; Processed kit ligand; SCF; SF; SHEP7; sKITLG; Sl; SLF; Soluble KIT ligand; Steel factor; steel factor/kit ligand; stem cell factor; stem cell factor KL-1; stem cell factor, CSF
Kitl
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.