missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ SCF (C-Kit Ligand) Recombinant Human Protein

Product Code. 11310842
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11310842

Brand: Gibco™ CTP2111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

SCF (C-Kit Ligand); Recombinant Human Protein

Cell Therapy Systems CTS™ Recombinant Human Stem Cell Factor (SCF) is a bioactive protein intended for use in cell culture applications, including pre-clinical and clinical cell therapy studies. SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development and hematopoiesis. These activities are all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis. The lyophilized, E. coli-expressed SCF CTS™ Recombinant Human is verified for activity through dose-dependent proliferation of human MO-7e cells.

  • Qualified—helps reduce your burden in qualifying reagents for regulatory submission in cell therapy applications
  • Highly Pure—minimizes interference from other proteins or contaminants
  • Active—get more results with less protein

Simplified Transition into Clinical Research

GIBCO™ CTS™ products are of high quality and supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The GIBCO™ CTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

High Purity

To help verify that GIBCO™ CTS™ Recombinant growth factors are of the highest quality, each protein is analyzed for purity, along with structural homogeneity, to help ensure a biologically active protein. The purity of each GIBCO™ CTS™ recombinant growth factor is verified by SDS-PAGE to be over 95% and to have endotoxin levels below 0.1 ng/μg.

High Biological Activity

All GIBCO™ CTS™ Recombinant growth factors are verified for activity. Each lot has its own ED50 determined using an activity assay based on a specific function of the growth factor. The activity of our SCF CTS™ Recombinant Human growth factor is determined by the dose-dependent proliferation of human MO-7e cells to have an ED50 range of 2 to 10 ng/mL (specific activity: 5 × 105–1 × 105 units/mg). Optimal concentration for individual applications should be determined by a dose response assay.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 18.5 kDa
Purification Method Sequential Chromatography
Quantity 100 μg
Source Human
Gene Alias SCF (c-kit Ligand)
Protein Length 164 aa
Species Human
Recombinant Recombinant
Content And Storage 2°C to 8°C
Classification Carrier-Free
Activity 2 ng/ml
Bioassay Proliferation of MO7e cells
Endotoxin Level <0.1 ng/μg
Shipping Condition Wet Ice
Expression System E. coli
Protein Family Growth Factors & Receptors
Protein Form Full Length, Recombinant, Ligand
Form Lyophilized
Protein Subtype Other Proteins
Research Category Clinical Research, Stem Cell Research, Differentiation, Oncology
Product Line Gibco
Purity or Quality Grade 95 %
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.