missing translation for 'onlineSavingsMsg'
Learn More

SCF/c-kit Ligand Antibody [DyLight 755], Novus Biologicals™

Product Code. 30734973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734973 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734973 Supplier Novus Biologicals Supplier No. AF6998Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SCF/c-kit Ligand Polyclonal antibody specifically detects SCF/c-kit Ligand in Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCF/c-kit Ligand
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Host Species Sheep
Immunogen E. coli-derived recombinant rat SCF/c-kit Ligand, Gln26-Ala189, Accession # P21581
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4254
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.