missing translation for 'onlineSavingsMsg'
Learn More

SCF/c-kit Ligand Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NBP1-77030-0.025mg

 View more versions of this product

Product Code. 18773254

  • £173.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SCF/c-kit Ligand Polyclonal specifically detects SCF/c-kit Ligand in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCF/c-kit Ligand
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor
Rabbit
23, 28 kDa
0.025 mg
Cancer, Embryonic Stem Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
4254
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL
P21583
KITLG
Antibody was raised against an 18 amino acid synthetic peptide from near the center of human SCF. The immunogen is located within amino acids 100 - 150 of SCF. Amino Acid Squence: KENSSKDLKKSFKSPEPR
Affinity Purified
RUO
Primary
Human, Mouse, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only