Learn More
Trajan Scientific and Medical™ SCeramic Capillary Tube Cutter
Click to view available options
For Use With (Equipment):
Fused Silica Tubing
Unit Size:
Pack of 3
Description
- Burr-free cutting of fused silica tubing
- Reduces likelihood of blockages
- Clean entry for on-column injection
Specifications
Specifications
| Type | Capillary Ceramic Tube Cutter |
| Quantity | 3/Pk. |
| For Use With (Equipment) | Fused Silica Tubing |