missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575757
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Stearoyl-CoA desaturase (SCD; EC 1.14.99.5) is an iron-containing enzyme that catalyzes a rate-limiting step in the synthesis of unsaturated fatty acids. The principal product of SCD is oleic acid, which is formed by desaturation of stearic acid. The ratio of stearic acid to oleic acid has been implicated in the regulation of cell growth and differentiation through effects on cell membrane fluidity and signal transduction. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5., have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.
Specifications
| SCD | |
| Polyclonal | |
| Unconjugated | |
| Scd | |
| AA589638; ab; acyl-CoA desaturase; Acyl-CoA desaturase 1; acyl-CoA desaturase 2; AI265570; asebia; delta(9)-desaturase; Delta(9)-desaturase 1; delta(9)-desaturase 2; Delta-9 desaturase; delta-9 desaturase 1; Delta-9 desaturase 2; delta-9-desaturase; FADS5; fatty acid desaturase; Fatty acid desaturase 1; fatty acid desaturase 2; HGNC:10571; hSCD1; MSTP008; OTTHUMP00000020280; predicted protein of HQ0998; PRO0998; PRO1933; Scd; Scd1; Scd-1; Scd2; SCDOS; stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); stearoyl-CoA desaturase 1; Stearoyl-CoA desaturase 2; stearoyl-CoA desaturase opposite strand; stearoyl-Coenzyme A desaturase 1; stearoyl-Coenzyme A desaturase 2; stearyl-CoA desaturase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20249, 246074, 6319 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00767, P07308, P13516 | |
| Scd, Scd1 | |
| Synthetic peptide corresponding to residues in Human SCD. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction