missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23393-1-AP-150UL
This item is not returnable.
View return policy
Description
Stearoyl-CoA desaturase., ,Specifications
| SCD | |
| Polyclonal | |
| Unconjugated | |
| SCD | |
| Acyl CoA desaturase, Delta(9) desaturase, FADS5, Fatty acid desaturase, MSTP008, SCD, SCD1, SCDOS, Stearoyl CoA desaturase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 246074, 6319 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00767 | |
| SCD | |
| SCD Fusion Protein Ag13456 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction