missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517331
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Stearoyl-CoA desaturase (SCD; EC 1.14.99.5) is an iron-containing enzyme that catalyzes a rate-limiting step in the synthesis of unsaturated fatty acids. The principal product of SCD is oleic acid, which is formed by desaturation of stearic acid. The ratio of stearic acid to oleic acid has been implicated in the regulation of cell growth and differentiation through effects on cell membrane fluidity and signal transduction. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 and SCD5., have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates.
Specifications
| SCD | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O00767, P13516 | |
| Scd, Scd1 | |
| Synthetic peptide corresponding to residues surrounding M38 of human SCD1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Scd | |
| AA589638; ab; acyl-CoA desaturase; Acyl-CoA desaturase 1; acyl-CoA desaturase 2; AI265570; asebia; delta(9)-desaturase; Delta(9)-desaturase 1; delta(9)-desaturase 2; Delta-9 desaturase; delta-9 desaturase 1; Delta-9 desaturase 2; delta-9-desaturase; FADS5; fatty acid desaturase; Fatty acid desaturase 1; fatty acid desaturase 2; HGNC:10571; hSCD1; MSTP008; OTTHUMP00000020280; predicted protein of HQ0998; PRO0998; PRO1933; Scd; Scd1; Scd-1; Scd2; SCDOS; stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); stearoyl-CoA desaturase 1; Stearoyl-CoA desaturase 2; stearoyl-CoA desaturase opposite strand; stearoyl-Coenzyme A desaturase 1; stearoyl-Coenzyme A desaturase 2; stearyl-CoA desaturase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20249, 6319 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction