missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nbs1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£169.00 - £395.00
Specifications
| Antigen | Nbs1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/mL. Use reported in scientific literature, Immunohistochemistry 2 ug/mL, Immunohistochemistry-Paraffin reported in scientific literature (PMID 21349997; 30176843) |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18127499
|
Novus Biologicals
NBP1-06609SS |
0.025 mL |
£169.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719103
|
Novus Biologicals
NBP1-06609 |
£395.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Nbs1 | |
| Western Blot 2 ug/mL. Use reported in scientific literature, Immunohistochemistry 2 ug/mL, Immunohistochemistry-Paraffin reported in scientific literature (PMID 21349997; 30176843) | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors | |
| PBS, 30% Glycerol with 0.1% Sodium Azide | |
| 4683 | |
| Nbs1 Antibody was made to a synthetic peptide made to an internal portion of the mouse NBS1 protein (within residues 350-400). [Swiss-Prot# Q9R207] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| NBN | |
| IgG | |
| Affinity Purified | |
| 84 kDa |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel