missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD-1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26393B
This item is not returnable.
View return policy
Description
SCD-1 Polyclonal specifically detects SCD-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SCD-1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| O00767 | |
| SCD | |
| Synthetic peptide made to an internal portion of human SCD (within residues 50-100). [Swiss-Prot# O00767] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| FADS5, MSTP008, SCD1, SCDOS, Stearoyl-CoA desaturase | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism, Signal Transduction, Stem Cell Markers | |
| 6319 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction