Learn More
Invitrogen™ SCCPDH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596070
Description
Immunogen sequence: MATEQRPFHL VVFGASGFTG QFVTEEVARE QVDPERSSRL PWAVAGRSRE KLQRVLEKAA LKLGRPTLSS EVGIIICDIA NPASLDEMAK QATVVLNCVG PYRFYGEPVI KACIENGASC IDISGEPQFL ELMQLKYHEK AADKGVYIIG SSGFDSIPAD LGVIYTRNKM NGTLTAVESF LTIHSGPEGL SIHDGTWKSA IYGFGDQSNL RKLRNVSNLK PVPLIGPKLK RRWPISYCRE LKGYSIPFMG SDVSVVRRTQ RYLYENLEES; Positive Samples: THP-1, PC-3, 22Rv1, U-251MG, A673, Mouse heart.
SCCPDH (Probable saccharopine dehydrogenase) is a 429 amino acid protein that belongs to the saccharopine dehydrogenase family. The SCCPDH gene is conserved in chimpanzee, dog, cow, mouse, rat, chicken, fruit fly, mosquito and C. elegans, and maps to human chromosome 1q44. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Stickler syndrome, Parkinson's, Gaucher disease and Usher syndrome are also associated with chromosome 1. A breakpoint has been identified in 1q which disrupts the DISC1 gene and is linked to schizophrenia. Aberrations in chromosome 1 are found in a variety of cancers including head and neck cancer, malignant melanoma and multiple myeloma.
Specifications
| SCCPDH | |
| Polyclonal | |
| Unconjugated | |
| SCCPDH | |
| AW214504; C330023F11Rik; CGI 49; CGI49; CGI-49; hypothetical protein LOC507289; NET11; probable saccharopine dehydrogenase; RGD1311440; RP11-439E19.2; saccharopine dehydrogenase (putative); saccharopine dehydrogenase-like oxidoreductase; Sccpdh; SCPDH | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109232, 51097 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.61 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NBX0, Q8R127 | |
| SCCPDH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SCCPDH (NP_0570862). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.