missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCCPDH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97630-100ul
This item is not returnable.
View return policy
Description
SCCPDH Polyclonal antibody specifically detects SCCPDH in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SCCPDH | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
| CGI-49, EC 1.5.1.9, FLJ43187, NET11, probable saccharopine dehydrogenase, RP11-439E19.2, saccharopine dehydrogenase (putative) | |
| Produced in rabbits immunized with E. coli-derived Human SCCPDH fragment. | |
| 100 μg | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 51097 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction