missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCCPDH Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97630CL1
This item is not returnable.
View return policy
Description
SCCPDH Polyclonal antibody specifically detects SCCPDH in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| SCCPDH | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| CGI-49, EC 1.5.1.9, FLJ43187, NET11, probable saccharopine dehydrogenase, RP11-439E19.2, saccharopine dehydrogenase (putative) | |
| Produced in rabbits immunized with E. coli-derived Human SCCPDH fragment. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 51097 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction