missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCARF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528426
This item is not returnable.
View return policy
Description
Recommended positive controls: HUVEC. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a scavenger receptor that is expressed in endothelial cells and mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). Transcription factors ZNF444/EZF-2 and SP1 are found to be responsible for the regulation of this gene. Five alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| SCARF1 | |
| Polyclonal | |
| Unconjugated | |
| Scarf1 | |
| AA986099; acetyl LDL receptor; endothelial cells scavenger receptor; KIAA0149; MGC47738; mKIAA0149; SCARF1; Scavenger receptor class F member 1; scavenger receptor class F, member 1; scavenger receptor expressed by endothelial cells 1; Scavenger receptor expressed by endothelial cells 1 protein; SREC; SREC1; SREC-I | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8578 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q14162 | |
| Scarf1 | |
| Synthetic peptide encompassing a sequence within the Extracellular domain of human SCARF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction