missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARA5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55390-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can recognize isoform 1,2 and 3 of SCARA5.
Scavenger receptor class A member 5 (SCARA5), also known as TESR, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARA5 is a cell-surface receptor that contains that collagen-like domain and N-terminal scavenger receptor cysteine rich domain typical of the class A scavenger receptor subfamily. Unlike other class A receptors, SCARA5 does not endocytose acetylated or oxidized low density lipoprotein. Instead, SCARA5 binds serum ferritin and mediates its endocytosis from the cell surface, indicating SCARA5 may play a role in non-transferrin iron delivery. At least four isoforms of SCARB1 are known to exist.Specifications
| SCARA5 | |
| Polyclonal | |
| Unconjugated | |
| SCARA5 | |
| NET33, SCARA5, Scavenger receptor hlg, Tesr | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 286133, 71145 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZMJ2, Q8K299 | |
| SCARA5 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto