missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCARA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523551
This item is not returnable.
View return policy
Description
SCARA5 Polyclonal Antibody for Western Blot, IHC (P)
Scavenger receptor class A member 5 (SCARA5), also known as TESR, is part of the scavenger receptor superfamily, which is composed of many members with diverse structures, expression patterns, and functions. SCARA5 is a cell-surface receptor that contains that collagen-like domain and N-terminal scavenger receptor cysteine rich domain typical of the class A scavenger receptor subfamily. Unlike other class A receptors, SCARA5 does not endocytose acetylated or oxidized low density lipoprotein. Instead, SCARA5 binds serum ferritin and mediates its endocytosis from the cell surface, indicating SCARA5 may play a role in non-transferrin iron delivery. At least four isoforms of SCARB1 are known to exist.
Specifications
| SCARA5 | |
| Polyclonal | |
| Unconjugated | |
| SCARA5 | |
| 4932433F15Rik; 4933425F03Rik; AV278087; NET33; SCARA5; Scavenger receptor class A member 5; scavenger receptor class A, member 5 (putative); scavenger receptor hlg; Tesr; testis expressed scavenger receptor; UNQ2938/PRO28700 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 286133, 71145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6ZMJ2, Q8K299 | |
| SCARA5 | |
| KLH conjugated synthetic peptide between 385-413 amino acids from the C-terminal region of human SCAR5. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction