missing translation for 'onlineSavingsMsg'
Learn More

SCARA5 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18412544 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412544 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412544 Supplier Novus Biologicals Supplier No. NBP177050AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCARA5 Polyclonal antibody specifically detects SCARA5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCARA5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias FLJ23907, MGC45780, scavenger receptor class A, member 5 (putative), testis expressed scavenger receptor, UNQ2938/PRO28700
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SCARA5. The immunogen is located within amino acids 350 - 400 of SCARA5. Amino Acid Squence: KGDRGPKGEKGEKGDRA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286133
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.