missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAP2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB110-40659
This item is not returnable.
View return policy
Description
SCAP2 Polyclonal specifically detects SCAP2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| SCAP2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Fyn-associated phosphoprotein SKAP55 homologue, PRAP, RA70Pyk2/RAFTK-associated protein, Retinoic acid-induced protein 70, SAPSSKAP55 homolog, SCAP2MGC33304, SKAP55RSKAP-55HOM, SKAP-HOMMGC10411, src family associated phosphoprotein 2, Src family-associated phosphoprotein 2, src kinase associated phosphoprotein 2, src kinase-associated phosphoprotein 2, Src kinase-associated phosphoprotein 55-related protein, src kinase-associated phosphoprotein of 55-related protein, Src-associated adapter protein with PH and SH3 domains, src-associated adaptor protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8935 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| O75563 | |
| SKAP2 | |
| Peptide with sequence C-GLVPKAYIMEMYDI corresponding to C-Terminus according to NP_003921.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction