missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAP Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04113APC
This item is not returnable.
View return policy
Description
SCAP Polyclonal antibody specifically detects SCAP in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| SCAP | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| KIAA0199, PSEC0227, SREBF chaperone, SREBP cleavage-activating protein, sterol regulatory element binding protein cleavage-activating protein | |
| Maps to a region between residue 450 and 500 of human Sterol Regulatory Element Binding Protein Ceavage-Activating Protein using the numbering given in entry NP_036367.2 (GeneID 22937). | |
| 0.1 mL | |
| Cardiovascular Biology, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction | |
| 22937 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction