missing translation for 'onlineSavingsMsg'
Learn More

Scandium Triflate, TRC

Product Code. 30419020
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419020 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419020 Supplier Toronto Research Chemicals Supplier No. TRCS19890025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Scandium Triflate
CAS 144026-79-9
Molecular Formula C3H3F9O9S3 . Sc
Quantity 25 g
Synonym Scandium(III) Triflate, Scandium(III) Trifluoromethanesulfonate, Scandium(3+) Triflate, Scandium Trifluoromethanesulfonate, Scandium Tris(trifluoromethanesulfonate), Trifluoromethanesulfonic Acid Scandium(3+) Salt
InChI Formula InChI=1S/3CHF3O3S.Sc/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3
SMILES [Sc+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
IUPAC Name scandium(3+);trifluoromethanesulfonate
Molecular Weight (g/mol) 492.2
Formula Weight 491.812
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.