missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) trifluoromethanesulfonate, 98%

Product Code. 11328548
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328548 5 g 5g
11400291 250 mg 250mg
11318548 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328548 Supplier Thermo Scientific Alfa Aesar Supplier No. 040566.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144026-79-9 | C3F9O9S3Sc | 492.146 g/mol

Scandium(III) trifluoromethanesulfonate is widely used as a catalyst in hydrothiolation, selective two-electron reduction of oxygen by ferrocene derivatives and vinylogous Fridel-crafts alkylation of indoles and pyrrole in water. It is involved in the Mukaiyama aldol addition and stereochemically catalyzes the radical polymerization of acrylates. It acts as a Lewis acid catalyst and used in the synthesis of bullvalone via a stabilized sulfur ylide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144026-79-9
Molecular Formula C3F9O9S3Sc
Molecular Weight (g/mol) 492.146
MDL Number MFCD00798539
InChI Key HZXJVDYQRYYYOR-UHFFFAOYSA-K
Synonym scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
PubChem CID 2734571
IUPAC Name scandium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]

Specifications

Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water,alcohol and acetonitrile.
Formula Weight 492.16
Percent Purity 98%
Sensitivity Hygroscopic
Chemical Name or Material Scandium(III) trifluoromethanesulfonate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.