missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Product Code. 11430311
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430311 2 g 2g
11440311 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430311 Supplier Thermo Scientific Alfa Aesar Supplier No. 040579.NA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14066-05-8
Molecular Formula Cl3O12Sc
Molecular Weight (g/mol) 343.294
MDL Number MFCD00243289
InChI Key UBVKRMCXDCYZQK-UHFFFAOYSA-K
Synonym scandium perchlorate, scandium 3+ perchlorate, acmc-20ajr3, scandium 3+ ion triperchlorate, trihyperchloric acid scandium salt, scandium iii perchlorate solution, scandium 3+ triperchlorate
PubChem CID 11013318
IUPAC Name scandium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Sc+3]

Specifications

Physical Form Liquid
Quantity 2 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 451.42 (343.30 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Scandium(III) perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.