missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) chloride hexahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11346268
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20662-14-0
Molecular Formula Cl3H12O6Sc
Molecular Weight (g/mol) 259.40
MDL Number MFCD00149852
InChI Key FKZFOHABAHJDIK-UHFFFAOYSA-K
IUPAC Name scandium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Sc+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
Merck Index 14,8392
Formula Weight 259.41 (151.32 Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Scandium(III) chloride hexahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'