missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number p-7049261
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
MFCD00011501
This item is not returnable. View return policy
MFCD00011501

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >500°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 269.94 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Scandium(III) carbonate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'