missing translation for 'onlineSavingsMsg'
Learn More

Scandium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11369288
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-20-2
Sc
44.96
MFCD00016323
SIXSYDAISGFNSX-UHFFFAOYSA-N
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
23952
CHEBI:33330
scandium
[Sc]
This item is not returnable. View return policy
7440-20-2
Sc
44.96
MFCD00016323
SIXSYDAISGFNSX-UHFFFAOYSA-N
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
23952
CHEBI:33330
scandium
[Sc]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Weight ≈0.20g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Scandium foil, 0.1mm (0.004 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'