missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP5 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3432CL1
This item is not returnable.
View return policy
Description
SCAMP5 Polyclonal antibody specifically detects SCAMP5 in Rat samples. It is validated for Western Blot
Specifications
| SCAMP5 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| DKFZp686L1799, hSCAMP5, MGC24969, Sc5, secretory carrier membrane protein 5, secretory carrier-associated membrane protein 5 | |
| Synthetic Peptide: C N(217) Q P Q T Q Y S A T P N Y T Y S N(233) | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 192683 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction