missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCAMP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521428
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, Molt-4. Predicted reactivity: Mouse (88%), Rat (83%), Pig (92%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene product belongs to the SCAMP family of proteins which are secretory carrier membrane proteins. They function as carriers to the cell surface in post-golgi recycling pathways. Different family members are highly related products of distinct genes, and are usually expressed together. These findings suggest that the SCAMPs may function at the same site during vesicular transport rather than in separate pathways. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| SCAMP3 | |
| Polyclonal | |
| Unconjugated | |
| SCAMP3 | |
| C1orf3; propin 1; PROPIN1; Sc3; scamp3; Secretory carrier membrane protein 3; secretory carrier membrane protein 3 isoform 1; secretory carrier-associated membrane protein 3; TU52; zgc:103488 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O14828 | |
| SCAMP3 | |
| Recombinant protein encompassing a sequence within the center region of human SCAMP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction