missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCAMP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553215
This item is not returnable.
View return policy
Description
Immunogen sequence: DPVDVNPFQD PSVTQLTNAP QGGLAEFNPF SETNAATTVP VTQLPGSSQP AVLQPSVEPT QPTPQAVVSA AQAGLLRQQE ELDRKAAELE RKERELQNTV ANLHV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Secretory carrier membrane proteins (SCAMPs) are proteins that are components of post-Golgi membranes. These proteins are implicated to function in membrane trafficking. In fibroblasts, SCAMPs are concentrated in compartments that are involved in the recycling of cell surface receptors and endocytosis. In neurons, SCAMPs are associated with synaptic vesicles, secretion granules and transporter vesicles. SCAMPs are composed of four central transmembrane regions and a cytoplasmic tail. Of the five known SCAMPs, SCAMPs 1-3 contain cytoplasmic N-terminal regions with NPF repeats. NPF repeats are found to interact with EH domain proteins that function in budding of transport vesicles from the plasma membrane or the Golgi complex. SCAMPs 4 and 5 lack the N-terminal NPF repeats. SCAMPs 1-4 are all ubiquitously coexpressed while SCAMP 5 is only detectable in the brain. Studies show that SCAMP 5 is expressed late in development which is coincident with expansion of mature synapses.
Specifications
| SCAMP2 | |
| Polyclonal | |
| Unconjugated | |
| Scamp2 | |
| AI875466; I79_023583; Sc2; scamp 2; SCAMP2; Secretory carrier membrane protein 2; secretory carrier-associated membrane protein 2; testis secretory sperm-binding protein Li 219p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10066, 24044, 65168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15127, Q9ERN0 | |
| Scamp2 | |
| Recombinant protein corresponding to Human SCAMP2. Recombinant protein control fragment (Product #RP-91153). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction