missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3431MFV610
This item is not returnable.
View return policy
Description
SCAMP2 Polyclonal antibody specifically detects SCAMP2 in Human, Mouse, Hamster samples. It is validated for Western Blot
Specifications
| SCAMP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| mFluor Violet 610 SE | |
| secretory carrier membrane protein 2secretory carrier-associated membrane protein 2 | |
| Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 10066 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction