missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCAMP2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
£421.00
Specifications
| Antigen | SCAMP2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | FITC |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18507050
|
Novus Biologicals
NB120-3431F |
0.1 mL | |||||||
Description
SCAMP2 Polyclonal antibody specifically detects SCAMP2 in Human, Mouse, Hamster samples. It is validated for Western BlotSpecifications
| SCAMP2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| secretory carrier membrane protein 2secretory carrier-associated membrane protein 2 | |
| Synthetic Peptide: M(1) S A F D T N P F A D P V D V N P(17) C | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| FITC | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS | |
| 10066 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title