missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Scafftag™ Polypropylene Chemtag Inserts

Product Code. 12932985
Change view
Click to view available options
Includes:
10 ChemTag inserts
50 ChemTag Inserts
Unit Size:
Pack of 10
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
12932985 10 ChemTag inserts Pack of 10
12942985 50 ChemTag Inserts Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12932985 Supplier Brady™ Supplier No. 806687
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Inserts for the Chemtag™ System. They contain all the pertinent information specific to the risks that hazardous substances pose, such as: substance type, details of hazard and precautions, supplier's name and emergency phone number, etc.

  • Contain all the pertinent information specific to the risks that hazardous substances pose, such as:
    • Substance type
    • Details of hazard and precautions
    • Supplier's name and emergency phone number

Specifications

Product Type Chemtag Insert
Alternate Part Number CHEM-GB-CTI-STD-10
Color Multicolor
Material Polypropylene
Displayed Text Chemical Details
For Use With (Application) Inspection tagging, Hazard communication Identification
Height (English) 7.6 in.
Height (Metric) 193 mm
Includes 10 ChemTag inserts
Quantity 1 Ea.
Temperature Range (Metric) -10°C to +90°C
Thickness 0.5 mm
Width (English) 5.67 in.
Width (Metric) 144 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.