missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Scafftag™ Polycarbonate Holder Polypropylene Insert Multitag™ Kit

Product Code. 12953005
Change view
Click to view available options
Material:
Polypropylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
12953005 Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12953005 Supplier Brady™ Supplier No. 806739
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Multitag™ is a complete equipment inspection/maintenance tagging system which can be used effectively on a wide range of equipment, ranging from machinery and mobile plant to contractors plant and equipment.

  • Multitag™ is a complete equipment inspection/maintenance tagging system which can be used effectively on a wide range of equipment, ranging from machinery and mobile plant to contractors plant and equipment.

Specifications

Product Type Multi-tag Kit
Alternate Part Number MUL-GB-EITH-L-A
Color Red on White
Material Polypropylene
For Use With (Application) Inspection tagging, Component and equipment labelling
Height (English) 8.27 in.
Height (Metric) 210 mm
Includes 10 Multitag Holders, 10 Multitag Inserts, 1 Pen
Quantity 1 Ea.
Thickness 0.5 mm
Width (English) 2.17 in.
Width (Metric) 55 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.