missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Scafftag™ Plastic A4 Permit Wallet

Product Code. 12963005
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
12963005 Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12963005 Supplier Brady™ Supplier No. 833760
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Scafftag™ Plastic A4 Permit Wallet for inspection tagging and lockout tagout.

  • Transparent Plastic

Specifications

Product Type A4 Permit Wallet
Alternate Part Number PW1
Color Transparent
Material Plastic
For Use With (Application) Inspection tagging, Lockout tagout
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.