missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCAF8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557334
This item is not returnable.
View return policy
Description
Immunogen sequence: LSMTPETVKD VGFGSLVIPG GSVASNLATS ALPAGNVFNA PTKQAEPEEK VPHLIDHQIS SGENTRSVIP NDISSNAAIL G Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
Regulates the p53 pathway to control the expansion growth of digestive organs. Target protein binds the repetitive C-terminal domain (CTD) of RNA polymerase II; may act with other CTD binding proteins to link transcription and pre-mRNA processing.
Specifications
| SCAF8 | |
| Polyclonal | |
| Unconjugated | |
| Scaf8 | |
| A630086M08Rik; A930036P18Rik; AA408306; AI447644; AI448652; AU015035; CCAP7; CDC5L complex-associated protein 7; CTD-binding SR-like protein rA8; fc50c06; fd99c08; KIAA1116; mKIAA1116; protein SCAF8; putative RNA-binding protein 16; RBM16; RNA binding motif protein 16; RNA binding motif protein 16-like; protein SCAF8; RNA-binding motif protein 16; SCAF8; SR-like CTD-associated factor 8; SR-related and CTD-associated factor 8; SR-related CTD associated factor 8; SR-related CTD-associated factor 8; wu:fc50c06; wu:fd99c08 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPN6 | |
| Scaf8 | |
| Recombinant protein corresponding to Human SCAF8. Recombinant protein control fragment (Product #RP-95057). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction