missing translation for 'onlineSavingsMsg'
Learn More

SC79, MedChemExpress

Product Code. 16423637
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16423637

Brand: Medchem Express HY18749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SC79, a unique specific and BBB permeable Akt activator, activates Akt in the cytosol and inhibits Akt membrane translocation. SC79 specifically binds to the PH domain of Akt.

SC79, a unique specific and BBB permeable Akt activator, activates Akt in the cytosol and inhibits Akt membrane translocation. SC79 specifically binds to the PH domain of Akt.

Specifications

Chemical Name or Material SC79
CAS 305834-79-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H17ClN2O5
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (274.14 mM; Need ultrasonic)
SMILES O=C(OCC)C(C#N)C1C(C(OCC)=O)=C(N)OC2=CC=C(Cl)C=C12
Molecular Weight (g/mol) 364.78
Formula Weight 364.78
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.