missing translation for 'onlineSavingsMsg'
Learn More

SC79, MedChemExpress

Product Code. 16421286
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421286 200 mg 200mg
16467618 10 mg 10mg
16460088 5 mg 5mg
16423637 50 mg 50mg
16417536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16421286 Supplier Medchem Express Supplier No. HY18749

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SC79, a unique specific and BBB permeable Akt activator, activates Akt in the cytosol and inhibits Akt membrane translocation. SC79 specifically binds to the PH domain of Akt.

SC79, a unique specific and BBB permeable Akt activator, activates Akt in the cytosol and inhibits Akt membrane translocation. SC79 specifically binds to the PH domain of Akt.

Specifications

Chemical Name or Material SC79
CAS 305834-79-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H17ClN2O5
Quantity 200 mg
Solubility Information DMSO : 100 mg/mL (274.14 mM; Need ultrasonic)
SMILES O=C(OCC)C(C#N)C1C(C(OCC)=O)=C(N)OC2=CC=C(Cl)C=C12
Molecular Weight (g/mol) 364.78
Formula Weight 364.78
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.