missing translation for 'onlineSavingsMsg'
Learn More

SC75741, TRC

Product Code. 30419857
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419857 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419857 Supplier Toronto Research Chemicals Supplier No. TRCS19992010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(6-Benzoyl-1H-benzimidazol-2-yl)-2-(1-thieno[3,2-d]pyrimidin-4-yl-4-piperidinyl)-4-thiazolecarboxamide
CAS 913822-46-5
Molecular Formula C29H23N7O2S2
Quantity 10 mg
Synonym SC75741, N-(6-Benzoyl-1H-benzimidazol-2-yl)-2-(1-thieno[3,2-d]pyrimidin-4-yl-4-piperidinyl)-4-thiazolecarboxamide
InChI Formula InChI=1S/C29H23N7O2S2/c37-24(17-4-2-1-3-5-17)19-6-7-20-22(14-19)34-29(33-20)35-27(38)23-15-40-28(32-23)18-8-11-36(12-9-18)26-25-21(10-13-39-25)30-16-31-26/h1-7,10,13-16,18H,8-9,11-12H2,(H2,33,34,35,38)
SMILES O=C(Nc1nc2ccc(cc2[nH]1)C(=O)c3ccccc3)c4csc(n4)C5CCN(CC5)c6ncnc7ccsc67
IUPAC Name N-(6-benzoyl-1H-benzimidazol-2-yl)-2-(1-thieno[3,2-d]pyrimidin-4-ylpiperidin-4-yl)-1,3-thiazole-4-carboxamide
Molecular Weight (g/mol) 565.67
Formula Weight 565.1355
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.