missing translation for 'onlineSavingsMsg'
Learn More

SC75741, MedChemExpress

Product Code. 16470357
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470357 50 mg 50mg
16446717 5 mg 5mg
16445006 100 mg 100mg
16424008 10 mg 10mg
16422936 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16470357 Supplier Medchem Express Supplier No. HY10496
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SC75741 is a broad and efficient NF-κB inhibitor with an IC50 of 200 nM for p65. SC75741 blocks influenza viruses (IV) replication. SC75741 impairs DNA binding of the NF-κB subunit p65, resulting in reduced expression of cytokines, chemokines, and pro-apoptotic factors. SC75741 subsequently inhibits caspase activation and blocks caspase-mediated nuclear export of viral ribonucleoproteins.

SC75741 is a broad and efficient NF-κB inhibitor with an IC50 of 200 nM for p65. SC75741 blocks influenza viruses (IV) replication. SC75741 impairs DNA binding of the NF-κB subunit p65, resulting in reduced expression of cytokines, chemokines, and pro-apoptotic factors. SC75741 subsequently inhibits caspase activation and blocks caspase-mediated nuclear export of viral ribonucleoproteins.

Specifications

Chemical Name or Material SC75741
CAS 913822-46-5
Color Brown
Purity Grade Notes Research
Molecular Formula C29H23N7O2S2
Quantity 50 mg
Solubility Information DMSO : 83.33 mg/mL (147.31 mM; Need ultrasonic)
SMILES O=C(NC1=NC(C=CC(C(C2=CC=CC=C2)=O)=C3)=C3N1)C4=CSC(C(CC5)CCN5C6=NC=NC7=C6SC=C7)=N4
Molecular Weight (g/mol) 565.67
Formula Weight 565.67
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.