missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SC65 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94640-0.02ml
This item is not returnable.
View return policy
Description
SC65 Polyclonal antibody specifically detects SC65 in Human, Rat samples. It is validated for Western Blot
Specifications
| SC65 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| leprecan-like 4, Leprecan-like protein 4, NO55, 55kDa, NOL55, Nucleolar autoantigen No55, SC65rat synaptonemal complex protein, synaptonemal complex protein SC65 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 338-437 of human P3H4 (NP_006446.1). FHRARWGLEEEDFQPREEAMLYHNQTAELRELLEFTHMYLQSDDEMELEETEPPLEPEDALSDAEFEGEGDYEEGMYADWWQEPDAKGDEAEAEPEPELA | |
| 0.02 mL | |
| Cell Cycle and Replication | |
| 10609 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction