missing translation for 'onlineSavingsMsg'
Learn More

SC57666, MedChemExpress

Product Code. p-200095921
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16483169

Brand: Medchem Express HYU00129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SC57666 is a selective COX2 inhibitor with an IC50 of 26 nM.

SC57666 is a selective COX2 inhibitor with an IC50 of 26 nM.

Specifications

Chemical Name or Material SC57666
CAS 158959-32-1
Color White
Purity Grade Notes Research
Molecular Formula C18H17FO2S
Quantity 1 mg
Solubility Information DMSO : ≥ 100 mg/mL (316.07 mM)
SMILES O=S(C1=CC=C(C2=C(C3=CC=C(F)C=C3)CCC2)C=C1)(C)=O
Molecular Weight (g/mol) 316.39
Formula Weight 316.39
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.