missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SC 79

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Activator of Akt; binds to the pleckstrin homology domain of Akt. Enhances Akt phosphorylation by upstream protein kinases; also enables cytosolic activation of Akt. Shown to suppress excitotoxicity-induced neuronal death in vitro and in vivo. Exhibits brain penetrance.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 364.78 |
| Purity | >99% |
| Molecular Weight (g/mol) | 364.782 |
| Chemical Name or Material | SC 79 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction