missing translation for 'onlineSavingsMsg'
Learn More

SC-514, MedChemExpress

Product Code. 16425868
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16425868 10 mg 10mg
16432017 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16425868 Supplier Medchem Express Supplier No. HY13802

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SC-514 is a selective IKK-2 inhibitor (IC50=11.2 μM), which does not inhibit other IKK isoforms or other serine-threonine and tyrosine kinases.

SC-514 is a selective IKK-2 inhibitor (IC50=11.2 μM), which does not inhibit other IKK isoforms or other serine-threonine and tyrosine kinases.

Specifications

Chemical Name or Material SC-514
Physical Form Solid
CAS 354812-17-2
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C9H8N2OS2
For Use With (Application) Cancer-Kinase/protease
Synonym GK 01140
Solubility Information DMSO : 100 mg/mL (445.83 mM; Need ultrasonic)
SMILES O=C(C1=C(N)C=C(C2=CSC=C2)S1)N
Molecular Weight (g/mol) 224.3
Formula Weight 224.3
Percent Purity 99.78%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.