missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SC 19220
GREENER_CHOICE

Product Code. 18005532
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18005532

Brand: Tocris Bioscience™ 1206/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective EP1 receptor antagonist

Selective EP1 receptor antagonist (IC50 = 6.7 μM for inhibition of [3H]-PGE2 binding to EP1 transfected COS cells).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19395-87-0
Molecular Formula C16H14ClN3O3

Specifications

Quantity 10 mg
Formula Weight 331.76
Purity >99%
Molecular Weight (g/mol) 331.756
Chemical Name or Material SC 19220
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.