missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SBNO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144246
This item is not returnable.
View return policy
Description
SBNO2 Polyclonal Antibody for ICC/IF, ELISA
Specifications
| SBNO2 | |
| Polyclonal | |
| Unconjugated | |
| SBNO2 | |
| KIAA0963; Protein strawberry notch homolog 2; SBNO2; SNO; STNO; strawberry notch homolog 2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 22904 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9Y2G9 | |
| SBNO2 | |
| Recombinant Human Protein strawberry notch homolog 2 protein (6-146AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction