missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SBNO1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25359-1-AP-150UL
This item is not returnable.
View return policy
Description
SBNO1, also named as Monocyte protein 3 or MOP3, is a 1393 amino acid protein, which belongs to the SBNO family. SBNO1 play an important role in development of the vertebrate central nervous system. SBNO1 has four isoforms with MW 154 kDa and 80 kDa.Specifications
| SBNO1 | |
| Polyclonal | |
| Unconjugated | |
| SBNO1 | |
| Monocyte protein 3, MOP 3, MOP3, SBNO1, Sno | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 243272, 55206 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A3KN83, Q689Z5 | |
| SBNO1 | |
| SBNO1 Fusion Protein Ag19005 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction