missing translation for 'onlineSavingsMsg'
Learn More

SbmA Antibody, Novus Biologicals™

Product Code. 18328073 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328073 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328073 Supplier Novus Biologicals Supplier No. NBP311267

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SbmA Polyclonal specifically detects SbmA in Bacteria samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SbmA
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunoprecipitation
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Peptide antibiotic transporter SbmA
Host Species Rabbit
Immunogen Recombinant SbmA
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 914529
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.