missing translation for 'onlineSavingsMsg'
Learn More

SBI-115, MedChemExpress

Product Code. 16480797
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480797 50 mg 50mg
16477207 5 mg 5mg
16475356 100 mg 100mg
16464488 10 mg 10mg
16413116 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480797 Supplier Medchem Express Supplier No. HY111534
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SBI-115 is a TGR5 (GPCR19) antagonist. SBI-115 decreases hepatic cystogenesis with polycystic liver diseases via inhibiting TGR5.

SBI-115 is a TGR5 (GPCR19) antagonist. SBI-115 decreases hepatic cystogenesis with polycystic liver diseases via inhibiting TGR5.

Specifications

Chemical Name or Material SBI-115
CAS 882366-16-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H13ClN2O4S
Quantity 50 mg
Solubility Information DMSO : 150 mg/mL (440.17 mM; Need ultrasonic)
SMILES O=C(C1=NC(S(=O)(CC)=O)=NC=C1Cl)OC2=CC=CC(C)=C2
Molecular Weight (g/mol) 340.78
Formula Weight 340.78
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.